2,4,6-三(十五氟庚基)-1,3,5-三嗪 ≥98.0% (通过GC)

供应商: TCI EUROPE

Synonyms: 2,4,6-三(全氟庚基)-1,3,5-三嗪

TCIAT0828-100MGEA 0 CNY
TCIAT0828-100MG
2,4,6-三(十五氟庚基)-1,3,5-三嗪 ≥98.0% (通过GC)
2,4,6-三(十五氟庚基)-1,3,5-三嗪
分子式: C₂₄F₄₅N₃
分子量: 1185,21 g/mol
MDL编号: MFCD00042439
CAS号: 21674-38-4
EINECS: 244-521-3

现在订购

产品技术规格

Appearance White to Almost white wax
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR