2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAT0859-0.1MLEA 0 CNY
TCIAT0859-0.1ML
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine ≥98.0% (通过GC)
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
分子式: C₁₂F₂₁N₃
分子量: 585,12 g/mol
沸点: 165°C
MDL编号: MFCD00042438
CAS号: 915-76-4
EINECS: 213-023-8

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR