3,5,5-三甲基己酰氯 ≥90.0% (通过GC)

供应商: TCI EUROPE
Danger

Synonyms: 异壬酰氯

TCIAT0951-25MLEA 0 CNY
TCIAT0951-25ML
3,5,5-三甲基己酰氯 ≥90.0% (通过GC)
3,5,5-三甲基己酰氯
分子式: C₉H₁₇ClO
分子量: 176,68 g/mol
沸点: 190°C
闪点: 140°C
MDL编号: MFCD00044028
CAS号: 36727-29-4
EINECS: 253-168-4
UN号: 3265
ADR: 8,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR