3-(Trifluoromethyl)phenyltrimethylammonium iodide ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAT1014-25GEA 445 CNY
TCIAT1014-25G
3-(Trifluoromethyl)phenyltrimethylammonium iodide ≥98.0% (通过滴定分析)
3-(Trifluoromethyl)phenyltrimethylammonium iodide
分子式: C₁₀H₁₃F₃IN
分子量: 331,12 g/mol
MDL编号: MFCD00059980
CAS号: 27389-57-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR