2,3,5,6-Tetrafluoro-1,4-phenylenediamine ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAT1110-1GEA 2025 CNY
TCIAT1110-1G
2,3,5,6-Tetrafluoro-1,4-phenylenediamine ≥98.0% (通过GC,滴定分析)
2,3,5,6-Tetrafluoro-1,4-phenylenediamine
分子式: C₆H₄F₄N₂
分子量: 180,11 g/mol
MDL编号: MFCD00039767
CAS号: 1198-64-7
EINECS: 214-837-6

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR