3-Nitrophenyl-p-toluenesulphonate ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAT1210-25GEA 0 CNY
TCIAT1210-25G
3-Nitrophenyl-p-toluenesulphonate ≥98.0% (通过GC)
3-Nitrophenyl-p-toluenesulphonate
分子式: C₁₃H₁₁NO₅S
分子量: 293,29 g/mol
MDL编号: MFCD00060140
CAS号: 3899-90-9

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 111.0 to 115.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR