11,11,12,12-Tetracyanonaphtho-2,6-quinodimethane ≥98.0% (以总氮计)
供应商: TCI EUROPE
|
Danger
|
TCIAT1246-100MGEA
0
CNY
TCIAT1246-100MG
11,11,12,12-Tetracyanonaphtho-2,6-quinodimethane ≥98.0% (以总氮计)
11,11,12,12-Tetracyanonaphtho-2,6-quinodimethane
分子式:
C₁₆H₆N₄ 分子量: 254,25 g/mol |
MDL编号:
MFCD00059115 CAS号: 6251-01-0 UN号: 3439 ADR: 6.1,III |
产品技术规格
Appearance | Dark red to Dark purple to Dark blue powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...