3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAT1247-10GEA 0 CNY
TCIAT1247-10G
3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (通过滴定分析)
3-(Trifluoroacetamido)pyrrolidine hydrochloride
分子式: C₆H₉F₃N₂O
分子量: 218,60 g/mol
MDL编号: MFCD01090944
CAS号: 84424-06-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR