3,4,5,11-Tetrahydroacenaphthene ≥96.0% (通过GC)

供应商: TCI EUROPE

TCIAT1249-10GEA 0 CNY
TCIAT1249-10G
3,4,5,11-Tetrahydroacenaphthene ≥96.0% (通过GC)
3,4,5,11-Tetrahydroacenaphthene
分子式: C₁₂H₁₄
分子量: 158,24 g/mol
MDL编号: MFCD00143054
CAS号: 26761-12-6
EINECS: 247-976-6

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR