4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone ≥96.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAT1286-5GEA 0 CNY
TCIAT1286-5G
4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone ≥96.0% (通过滴定分析)
4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone
分子式: C₁₂H₉F₃N₂O₂
分子量: 270,21 g/mol
MDL编号: MFCD00143378
CAS号: 1691-93-6

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 96.0 %
Melting point 144.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR