1,2,3-Triphenylguanidine ≥97.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAT1316-25GEA 0 CNY
TCIAT1316-25G
1,2,3-Triphenylguanidine ≥97.0% (通过HPLC,滴定分析)
1,2,3-Triphenylguanidine
分子式: C₁₉H₁₇N₃
分子量: 287,37 g/mol
MDL编号: MFCD00020648
CAS号: 101-01-9
EINECS: 202-907-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 144.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR