3-噻吩丙二酸 ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
α-羧基噻吩-3-乙酸
, (3-噻吩基)丙二酸
TCIAT1354-25GEA
2050
CNY
TCIAT1354-25G
TCIAT1354-5G
3-噻吩丙二酸 ≥98.0% (通过HPLC,滴定分析)
3-噻吩丙二酸
分子式:
C₇H₆O₄S 分子量: 186,18 g/mol |
MDL编号:
MFCD00005469 CAS号: 21080-92-2 EINECS: 244-198-9 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...