1,1,1-Trichloroacetone ≥95.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAT1376-25GEA 0 CNY
TCIAT1376-25G
1,1,1-Trichloroacetone ≥95.0% (通过GC)
1,1,1-Trichloroacetone
分子式: C₃H₃Cl₃O
分子量: 161,41 g/mol
沸点: 134°C
闪点: 64°C
MDL编号: MFCD00018829
CAS号: 918-00-3

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR