2,4,6-Triphenylpyrylium hydrogensulphate ≥97.0% (通过HPLC)

供应商: TCI EUROPE
Danger

TCIAT1445-1GEA 0 CNY
TCIAT1445-1G
2,4,6-Triphenylpyrylium hydrogensulphate ≥97.0% (通过HPLC)
2,4,6-Triphenylpyrylium hydrogensulphate
分子式: C₂₃H₁₈O₅S
分子量: 406,45 g/mol
MDL编号: MFCD00012002
CAS号: 51071-75-1
UN号: 1759
ADR: 8,III

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR