1,2,4-Tris(methanesulphonyloxy)butane

供应商: TCI EUROPE

TCIAT1454-25GEA 950 CNY
TCIAT1454-25G
1,2,4-Tris(methanesulphonyloxy)butane
1,2,4-Tris(methanesulphonyloxy)butane
分子式: C₇H₁₆O₉S₃
MDL编号: MFCD00191688
CAS号: 108963-16-2

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Elemental analysis (Sulfuride) 27.70 to 28.80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR