2-Ethoxyethyl trichloroacetate ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAT1474-25GEA 590 CNY
TCIAT1474-25G
2-Ethoxyethyl trichloroacetate ≥95.0% (通过GC)
2-Ethoxyethyl trichloroacetate
分子式: C₆H₉Cl₃O₃
分子量: 235,49 g/mol
MDL编号: MFCD00191691
CAS号: 30668-97-4

现在订购

产品技术规格

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR