(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol

供应商: TCI EUROPE

TCIAT1482-1GEA 0 CNY
TCIAT1482-1G
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol
(S)-(-)-1,1,2-三苯基-1,2-乙二醇
分子式: C₂₀H₁₈O₂
分子量: 290,36 g/mol
MDL编号: MFCD00075492
CAS号: 108998-83-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 126.0 to 130.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR