2,4,5,6-Tetramethylbenzenedisulphonyl dichloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAT1562-5GEA
0
CNY
TCIAT1562-5G
2,4,5,6-Tetramethylbenzenedisulphonyl dichloride ≥98.0% (通过滴定分析)
2,4,5,6-Tetramethylbenzenedisulphonyl dichloride
分子式:
C₁₀H₁₂Cl₂O₄S₂ 分子量: 331,22 g/mol |
MDL编号:
MFCD00191502 CAS号: 97997-76-7 UN号: 3261 ADR: 8,III |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...