3,3,3′,3′-四甲基-1,1′-螺旋联吲哚-5,5′,6,6′-四醇 ≥96.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAT1737-25GEA 0 CNY
TCIAT1737-25G
3,3,3′,3′-四甲基-1,1′-螺旋联吲哚-5,5′,6,6′-四醇 ≥96.0% (通过HPLC,滴定分析)
3,3,3′,3′-四甲基-1,1′-螺旋联吲哚-5,5′,6,6′-四醇
分子式: C₂₁H₂₄O₄
分子量: 340,42 g/mol
MDL编号: MFCD00021235
CAS号: 77-08-7
EINECS: 201-003-1

现在订购

产品技术规格

Appearance White to Green to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR