1,5,9-三氮杂环十二烷 ≥95.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAT1876-100MGEA 0 CNY
TCIAT1876-100MG
1,5,9-三氮杂环十二烷 ≥95.0% (通过GC)
1,5,9-三氮杂环十二烷
分子式: C₉H₂₁N₃
分子量: 171,29 g/mol
CAS号: 294-80-4
UN号: 3259
ADR: 8,II

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR