2,3,4,6-四-O-苄基-D-吡喃葡萄糖酰氟,主要为 α 构型 ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAT1922-500MGEA 0 CNY
TCIAT1922-500MG
2,3,4,6-四-O-苄基-D-吡喃葡萄糖酰氟,主要为 α 构型 ≥95.0% (通过GC)
2,3,4,6-四-O-苄基-D-吡喃葡萄糖酰氟,主要为 α 构型
分子式: C₃₄H₃₅FO₅
分子量: 542,65 g/mol
存储条件: Refrigerator
MDL编号: MFCD03701104
CAS号: 89025-46-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 67.0 to 71.0 deg-C
Specific rotation [a]20/D +2.0 to +4.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR