2,3,4,6-Tetra-O-benzyl-β-D-glucopyranosyl fluoride ≥96.0% (通过HPLC)

供应商: TCI EUROPE

TCIAT1923-500MGEA 1025 CNY
TCIAT1923-500MG
2,3,4,6-Tetra-O-benzyl-β-D-glucopyranosyl fluoride ≥96.0% (通过HPLC)
2,3,4,6-Tetra-O-benzyl-beta-D-glucopyranosyl fluoride
分子式: C₃₄H₃₅FO₅
存储条件: Refrigerator
MDL编号: MFCD01862264
CAS号: 78153-79-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 45.0 to 49.0 deg-C
Specific rotation [a]20/D +29.0 to +33.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR