3,4,6-Tri-O-benzyl-2-deoxy-D-galactopyranose ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAT1932-100MGEA 905 CNY
TCIAT1932-100MG
3,4,6-Tri-O-benzyl-2-deoxy-D-galactopyranose ≥98.0% (通过HPLC)
3,4,6-Tri-O-benzyl-2-deoxy-D-galactopyranose
分子式: C₂₇H₃₀O₅
存储条件: Refrigerator
MDL编号: MFCD06797169
CAS号: 94189-64-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 83.0 to 87.0 deg-C
Specific rotation [a]20/D +39.0 to +44.0 deg (C=1, Chloroform) (24hrs allow)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR