6-O-(三异丙基硅基)-D-葡萄烯糖 ≥95.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAT1936-100MGEA 0 CNY
TCIAT1936-100MG
6-O-(三异丙基硅基)-D-葡萄烯糖 ≥95.0% (通过GC)
6-O-(三异丙基硅基)-D-葡萄烯糖
分子式: C₁₅H₃₀O₄Si
分子量: 302,49 g/mol
CAS号: 137915-37-8

现在订购

产品技术规格

Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR