2,4,6-Triisopropylbenzyl chloride ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAT1989-5GEA 0 CNY
TCIAT1989-5G
2,4,6-Triisopropylbenzyl chloride ≥98.0% (通过GC)
2,4,6-Triisopropylbenzyl chloride
分子式: C₁₆H₂₅Cl
分子量: 252,83 g/mol
存储条件: Refrigerator
MDL编号: MFCD03701106
CAS号: 38580-86-8
UN号: 3261
ADR: 8,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 34.0 to 37.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR