9H,9H-Triacontafluoro-8,10-heptadecanedione ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAT2037-100MGEA
0
CNY
TCIAT2037-100MG
9H,9H-Triacontafluoro-8,10-heptadecanedione ≥98.0% (通过GC)
9H,9H-Triacontafluoro-8,10-heptadecanedione
分子式:
C₁₇H₂F₃₀O₂ 分子量: 808,15 g/mol |
CAS号:
36554-97-9 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...