4,4',4''-Tris[phenyl(m-tolyl)amino]triphenylamine ≥98.0% (以总氮计)

供应商: TCI EUROPE

TCIAT2251-100MGEA 0 CNY
TCIAT2251-100MG
4,4',4''-Tris[phenyl(m-tolyl)amino]triphenylamine ≥98.0% (以总氮计)
4,4',4''-Tris[phenyl(m-tolyl)amino]triphenylamine
分子式: C₅₇H₄₈N₄
分子量: 789,04 g/mol
溶点: 210°C
MDL编号: MFCD00799401
CAS号: 124729-98-2

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR