2,3,4-Trihydroxydiphenylmethane ≥98.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
TCIAT2259-5GEA
0
CNY
TCIAT2259-5G
2,3,4-Trihydroxydiphenylmethane ≥98.0% (通过HPLC)
2,3,4-Trihydroxydiphenylmethane
分子式:
C₁₃H₁₂O₃ 分子量: 216,24 g/mol 沸点: 255°C |
CAS号:
17345-66-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 94.0 to 98.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...