2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture)

供应商: TCI EUROPE

TCIAT2279-5GEA 0 CNY
TCIAT2279-5G
2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture)
2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture)
分子式: C₁₂H₂₆O
分子量: 186,34 g/mol
沸点: 113°C
MDL编号: MFCD00026491
CAS号: 123-17-1
EINECS: 204-606-8

现在订购

产品技术规格

Appearance Colorless to Almost colorless powder to crystal
Purity (GC) min. 90.0 % (threo+erythro)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR