2,3,4,5-Tetrachlorobenzoyl chloride ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAT2379-25GEA 0 CNY
TCIAT2379-25G
2,3,4,5-Tetrachlorobenzoyl chloride ≥98.0% (通过GC,滴定分析)
2,3,4,5-Tetrachlorobenzoyl chloride
分子式: C₇HCl₅O
分子量: 278,33 g/mol
CAS号: 42221-52-3
EINECS: 406-760-3
UN号: 3261
ADR: 8,II

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR