1,3,5-Tris[4-(trifluoromethanesulphonyloxy)-3-(trimethylsilyl)phenyl]benzene ≥92.0% (通过HPLC)

供应商: TCI EUROPE
Danger

TCIAT2467-1GEA 0 CNY
TCIAT2467-1G
1,3,5-Tris[4-(trifluoromethanesulphonyloxy)-3-(trimethylsilyl)phenyl]benzene ≥92.0% (通过HPLC)
1,3,5-Tris[4-(trifluoromethanesulphonyloxy)-3-(trimethylsilyl)phenyl]benzene
分子式: C₃₆H₃₉F₉O₉S₃Si₃
CAS号: 847925-63-7
UN号: 3261
ADR: 8,II

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 92.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR