(S)-(+)-四氢糠胺 ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAT2552-1GEA
1050
CNY
TCIAT2552-1G
TCIAT2552-200MG
(S)-(+)-四氢糠胺 ≥98.0% (通过GC,滴定分析)
(S)-(+)-四氢糠胺
分子式:
C₅H₁₁NO 分子量: 101,15 g/mol 沸点: 52°C 闪点: 46°C |
MDL编号:
MFCD00085303 CAS号: 7175-81-7 UN号: 2943 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +13.0 to +18.0 deg (C=2, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...