2,3,4,6-Tetra-O-pivaloyl-D-mannopyranosyl fluoride ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAT2568-1GEA 0 CNY
TCIAT2568-1G
2,3,4,6-Tetra-O-pivaloyl-D-mannopyranosyl fluoride ≥95.0% (通过GC)
2,3,4,6-Tetra-O-pivaloyl-D-mannopyranosyl fluoride
分子式: C₂₆H₄₃FO₉
分子量: 518,62 g/mol
存储条件: Refrigerator
MDL编号: MFCD11035863
CAS号: 187269-63-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR