4-(Trimethylsilyl)diphenylacetylene ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAT2596-5GEA 0 CNY
TCIAT2596-5G
4-(Trimethylsilyl)diphenylacetylene ≥97.0% (通过GC)
4-(Trimethylsilyl)diphenylacetylene
分子式: C₁₇H₁₈Si
分子量: 250,42 g/mol
存储条件: Refrigerator
MDL编号: MFCD09953470
CAS号: 136459-72-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 37.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR