2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAT2605-25GEA 0 CNY
TCIAT2605-25G
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) ≥98.0% (通过GC)
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers)
分子式: C₁₀H₁₈O₂
分子量: 170,25 g/mol
沸点: 98°C
闪点: 100°C
MDL编号: MFCD06200770
CAS号: 14049-11-7
EINECS: 237-889-1

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR