2,2':5',2''-Terthiophene-5,5''-dicarboxaldehyde ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAT2639-1GEA
0
CNY
TCIAT2639-1G
2,2':5',2''-Terthiophene-5,5''-dicarboxaldehyde ≥98.0% (通过GC)
2,2':5',2''-Terthiophene-5,5''-dicarboxaldehyde
分子式:
C₁₄H₈O₂S₃ 分子量: 304,40 g/mol |
CAS号:
13130-50-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...