1-十一烷硫醇 ≥96.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 1-UDT11-巯基-1-十一醇十一烷基硫醇

TCIAU0066-5MLEA 0 CNY
TCIAU0066-5ML
1-十一烷硫醇 ≥96.0% (通过GC)
1-十一烷硫醇
分子式: C₁₁H₂₄S
分子量: 188,37 g/mol
沸点: 104°C
溶点: -3°C
闪点: 109°C
MDL编号: MFCD00022097
CAS号: 5332-52-5

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR