Disposable particle filtering respirator, FFP2, standard comfort series, Makrite®
COVTFP2SLWEA 1199 CNY
COVTFP2SLW
Disposable particle filtering respirator, FFP2, standard comfort series, Makrite®
呼吸器 一次性防护口罩 Particulate Respirators
Makrite® Standard Comfort series masks are filtering half-masks designed to protect against particle intrusion.
  • Excellent filter material for low breathing resistance
  • Individually adjustable plastic concealed nose clip for contoured fit
  • Ultrasonic welded head-straps for safe and secure seal
  • Can be easily paired with safety goggles, helmets, glasses or earmuffs
A properly selected respirator is essential to protect the wearer's health. Consult a safety professional to determine the suitability for intended applications. All materials are latex-free.
Colour: White.
Certifications: EN 149:2001+A1:2009