Double spatulas
供应商: Corning®
Corning®
CORN3003EA 2625 CNY
CORN3003 CORN3005 CORN3007 CORN3004 CORN3006
Double spatulas
样品勺
PS, white. Available in five different configurations, Corning® spatulas are designed to save time and to provide contamination-free samples.
  • Individually packaged
  • Certified RNase- and DNase-free, non pyrogenic, anti-static and sterile
  • Eliminates the potential for cross-contamination and resterilising necessary with reusable spatulas