Water bath disinfection reagent, Aquabator-Clean™
供应商: PanReac AppliChem
APLIA9390.0250EA 0 CNY
APLIA9390.0250
Water bath disinfection reagent, Aquabator-Clean™
清洗剂 水浴保护剂
Aquabator-Clean™ is designed for disinfecting water baths. It protects against growth of fungi and bacteria.
  • Supplied as a concentrate - 10 ml per litre of water
  • Biodegradable
  • Non irrtating to skin at recommended concentrations
Caution: Use biocides safely. Always read the label and product information before use.
Not suitable for CO₂ incubators.