Blotting membranes, Amersham™ Hybond™-N
供应商: Cytiva
GEHERPN303NEA 0 CNY
GEHERPN303N
Blotting membranes, Amersham™ Hybond™-N
Amersham™ Hybond™-N is a neutral nylon membrane with a binding capacity for nucleic acids up to 600 µg/cm².
  • Recommended for nucleic acid blotting and microarray applications
Amersham™ Hybond™-N is a strong, supported membrane that is inherently hydrophilic and requires no pre-wetting. Nucleic acids can be quickly and easily crosslinked to nylon using UV light. When using low buffer volumes Hybond™-NX is recommended.
Pore size: 0,45 µm
Nucleic acid size required: >50 bp