Order Entry
Protease inhibitor cocktail, Halt
Protease inhibitor cocktail, Halt
Certificates
undefined
Protease inhibitor cocktail, Halt
Specifications
  • Source:
  • Species:
  • Environmentally Preferable:
Specifications
Product Information
  • Catalog #
    Supplier #
    Form
    Pk
    Availability
    Price
  • Halt protease inhibitor cocktail (100X), sufficient to treat 1 L of sample
    Halt protease inhibitor cocktail (100X), sufficient to treat 1 L of sample
    PIER78438
    78438
    Liquid
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 毫升
    1 * 10 毫升 - ¥0.00
    ¥0.00
    1 * 10 毫升
       ¥0.00
  • Halt protease inhibitor cocktail, EDTA-free (100X), 1 ml, sufficient to treat 100 ml of sample
    Halt protease inhibitor cocktail, EDTA-free (100X), 1 ml, sufficient to treat 100 ml of sample
    PIER87785
    87785
    Liquid
    1
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥0.00