Engineered for durable, reliable and energy efficient operation, these Glassware Washers can be either Freestanding or Mobile and are supplied with mounting feet and wheels.
- 304 stainless steel washing chamber and full exterior casing ensures long life and corrosion protection plus maintains superior cosmetic appearance
- Three separate filters protect pumps from debris and are conveniently located for easy access
- Multiple cycle selections and microprocessor control systems ensure that each load is processed efficiently with minimum energy consumption
- Enhanced dual fan drying system
- Temperature selection to 70 °C (158 °F) for optimal cleaning of different loads
- Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy cost
- Spindle Rack Ready models can accommodate 1 or 2 direct injection spindle racks
Designed for simple operation and flexible loading, they can accommodate a wide range of requirements. New improved units feature a soft touch controller, improved drying, a quieter run, and are compatible with existing racks and accessories. Choose from three different models: Standard Freestanding and Mobile Washer, Freestanding and Mobile Washer with DI Rinse, or Freestanding and Mobile Washer with DI Rinse, Spindle Rack Ready.
Certifications: ETL and ETL-C listed.
订购须知 A wide range of accessories are available to fit both upper and lower racks; spindle racks, specialty baskets, and other inserts. For more information, contact your Avantor representative.