Scoop type spatula
供应商: VWR, part of Avantor
VWR®
VWRU470149-438EA 422.28 CNY
VWRU470149-438
Scoop type spatula
样品勺
This scoop-type spatula is versatile and durable.
  • Semicircular cross section
  • Stainless steel construction
  • Spatula dimensions: ¹/₂×7¹/₂"
It also has one rounded end and one pointed end.