VWR®, Erlenmeyer Flask, Narrow Mouth
供应商: VWR
VWR®
VWRU10545-846EA 1815.35 CNY
VWRU10545-846
VWR®, Erlenmeyer Flask, Narrow Mouth
烧瓶 三角烧瓶
Flask is durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.
  • Borosilicate glass fabrication
  • Durable design
  • Graduation range 1000 to 4000 ml
  • Rubber stopper with stopper size: 10
Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.