VWR®, PCR Plates, 96-Well, Thermal Cycling
供应商: VWR
VWR®
VWRU89049-178EA 599.33 CNY
VWRU89049-178
VWR®, PCR Plates, 96-Well, Thermal Cycling
微孔板 PCR板
Plates feature a smooth rim that allows better contact inside the block to ensure a perfect fit and easy removal, as well as printed alphanumeric labeling for easy well location.
  • Inert polypropylene construction
  • Each well holds 200 µl
Designed to fit ABI PRISM® 3700 series thermocyclers.