PIG® 4-in-1® Mat
NEWPMAT284
: NEW PIG
- Description:Mat
- L×W:24 m×410 mm
- Seq:02
- Environmentally Preferable:
- Absorbency:34,8 L
- Packed:1 roll/dispenser
- Pk:1
This versatile mat roll can also do work as a pad, sock or wipes. You can stock just one absorbent and still handle a variety of leaks, drips and spills.
- Exclusive 4-in-1 mat is a pad, roll, sock and wipes all in one
- Use as a pad to catch drips and soak up spills; roll out to cover large areas; fold along score lines to make a thick, absorbent "sock"; tear off a piece to use as a low-lint wipes
- No matter how you use it, this mat builds on the proven eight-layer, 100% PP construction of PIG Mat
- Scuff-resistant, spunbond outer layer resists wear and tear, even under heavy foot traffic or abrasive conditions
- Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
- Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
- Absorbs and retains oils, coolants, solvents and water
The flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job. Dispenser box makes mat easy to access, carry and store whilst protecting contents from dirt and moisture. Rolls are perforated every 25cm; easy to tear to size, use only what you need. 25% or more recycled content.