Pipetting robot, ASSIST PLUS
供应商: Integra Biosciences
ASSIST PLUS
INTL4505EA 0 CNY
INTL4505
Pipetting robot, ASSIST PLUS
移液管和移液器 重复移液管
INTEGRA has developed the ASSIST PLUS pipetting robot to streamline routine pipetting tasks. Using INTEGRA electronic multichannel pipettes, the system automates pipetting tasks, eliminates physical strain and ensures superior reproducibility and error free pipetting.
  • Automates any VOYAGER or VIAFLO multichannel electronic pipette
  • Compatible with all INTEGRA tube racks, reagent reservoirs and microplates (12-384 well)
  • Protocols are created either on the pipette or in VIALAB (free for download)
  • Easy and fast programming with the VIALAB pipetting software
Simply mount an INTEGRA VIAFLO or VOYAGER electronic pipette onto the ASSIST PLUS pipette holder, choose a pipetting protocol, add a tip rack and your labware. Press RUN and ASSIST PLUS will automatically carry out the pipetting task. ASSIST PLUS can be used for serial dilutions, reformatting, plate filling and reagent addition and it’s possible to add options of heating, cooling and shaking of the labware. For setting up the protocol you use VIALAB – an intuitive computer program where you can visualize the protocol before running it on the ASSIST PLUS.
订购须知 For a fully functional ASSIST PLUS you need the base unit (INTL4505), a suitable electronic pipette (VIAFLO or VOYAGER) with a Bluetooth communication module installed and a charger stand. In addition you would need tips and suitable labware for your experiment. Please contact us for an offer based on your needs.
Delivery information: Includes base unit, 3 position universal deck, waste bin with frame and reflector, pipette charging cable, power cord and USB cable.