Developed to provide comprehensive centrifugation solutions. This micro-centrifuge features a powerful brushless DC motor and an enhanced airflow design to protect sensitive samples from rising temperatures. Additional features such as imbalance detection, lid lock safety and one-touch toggle make it very easy and convenient to use.
- Microprocessor controlled centrifuge with variable speed and time setting, along with last run memory function
- One touch short spin operation and change over from RPM to RCF
- Imbalance detection feature for increased safety
- Better airflow design ensures less heat generation, greater sample safety and less noise
- Lid lock safety- Lid opens automatically when run is complete
- Small footprint saves valuable bench space
- Suitable for use in fume hoods or cold room
Delivery information: Supplied with rotor as standard.