VWR®, High Speed Microcentrifuge
供应商: VWR
VWR®
VWRA521-2797EA 14034.24 CNY
VWRA521-2797
VWR®, High Speed Microcentrifuge
离心机 微型离心机
Developed to provide comprehensive centrifugation solutions. This micro-centrifuge features a powerful brushless DC motor and an enhanced airflow design to protect sensitive samples from rising temperatures. Additional features such as imbalance detection, lid lock safety and one-touch toggle make it very easy and convenient to use.
  • Microprocessor controlled centrifuge with variable speed and time setting, along with last run memory function
  • One touch short spin operation and change over from RPM to RCF
  • Imbalance detection feature for increased safety
  • Better airflow design ensures less heat generation, greater sample safety and less noise
  • Lid lock safety- Lid opens automatically when run is complete
  • Small footprint saves valuable bench space
  • Suitable for use in fume hoods or cold room
Delivery information: Supplied with rotor as standard.